missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Apoferritin from Horse Spleen

Protein shell of ferritin, abundant in beta-cells of pancreas

Supplier:  MP Biomedicals™ 0210026090

 View more versions of this product

Catalog No. ICN10026090


Only null left
Explore available promotions

Description

Description

When added to cultured endothelial cells apoferritin is taken up in a dose-responsive manner and appears to protect the cells from oxidant-mediated cytolysis.

  • CAS Number: 9013-31-4
  • MDL Number: MFCD00081365
  • Format: Off-white lyophilized powder
  • Protein Content: ≈90%
  • Iron Content: Typically <0.01%
  • Soluble in water or dilute buffer

Used for calibration of gel filtration columns and sodium dodecyl sulfate (SDS)-polyacrylamide gel electrophoresis. It acts as an anti-oxidant.

Specifications

Specifications

Molecular Biology
Apoferritin from Horse Spleen
Horse Spleen
Lyophilized
445000
500 mg
-20°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions