missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Spectinomycin, dihydrochloride pentahydrate, PhytoPure™

An aminocyclitol antibiotic, closely related to the aminoglycosides, it inhibits protein synthesis (elongation) by interfering with peptidyl tRNA translocation. It is active against gram-negative bacteria and is used as a selection agent for transformed plant cells that contain the selectable marker gene Spcr.

Supplier:  MP Biomedicals™ 0219454025

 View more versions of this product

Catalog No. ICN19454025


Only null left
Explore available promotions

Description

Description

 

  • Format: Crystalline Powder, Non-sterile
  • Synonyms: U18409AE; Actinospectacin; M141; Trobicin  

Specinomycin is used in cell culture applications, It is active against gram-negative bacteria.It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation systems for plant cells containing the marker gene Spcr, to study respiratory tract infections of cattle caused by Pasteurella multocida and Mannheimia haemolytica, and to generate plants deficient for the plastid-encoded RNA polymerase on MS-agar media. Spectinomycin also has use in amplification of low copy number plasmid carrying replicons.

All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.

Specifications

Specifications

22189-32-8
Crystalline powder
25 g
4°C
Antibiotic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions