Learn More
MP Biomedicals™ Spectinomycin, dihydrochloride pentahydrate, PhytoPure™
An aminocyclitol antibiotic, closely related to the aminoglycosides, it inhibits protein synthesis (elongation) by interfering with peptidyl tRNA translocation. It is active against gram-negative bacteria and is used as a selection agent for transformed plant cells that contain the selectable marker gene Spcr.
$169.67 - $677.81
Specifications
| CAS | 22189-32-8 |
|---|---|
| Content And Storage | 4°C |
| Form | Crystalline powder |
| Product Type | Antibiotic |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN19454005
|
MP Biomedicals™
0219454005 |
5 g |
Each for $169.67
|
|
|||||
|
ICN19454025
|
MP Biomedicals™
0219454025 |
25 g |
Each for $677.81
|
|
|||||
Description
- Format: Crystalline Powder, Non-sterile
- Synonyms: U18409AE; Actinospectacin; M141; Trobicin
Specinomycin is used in cell culture applications, It is active against gram-negative bacteria.It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation systems for plant cells containing the marker gene Spcr, to study respiratory tract infections of cattle caused by Pasteurella multocida and Mannheimia haemolytica, and to generate plants deficient for the plastid-encoded RNA polymerase on MS-agar media. Spectinomycin also has use in amplification of low copy number plasmid carrying replicons.
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Specifications
| 22189-32-8 | |
| 4°C | |
| Crystalline powder | |
| Antibiotic |