missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Alpha-Amylase

Catalog No. ICN10044725
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10044725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10044725 Supplier MP Biomedicals™ Supplier No. 0210044725

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

From Bacillus subtilis

  • CAS Number: 9000-90-2
  • EC Number: 232-565-6
  • Activity: 500-1,000 units per mg protein

Specifications

Color Off-White
pH 5 to 7
For Use With (Application) General procedure for the enzymatic liquefaction
Content And Storage 4°C
Source Bacillus subtilis
Quantity 25 g
Product Type alpha-Amylase
Form Powder