missing translation for 'onlineSavingsMsg'
Learn More

α-Amylase from Porcine pancreas, MP Biomedicals™

Catalog No. MP219123910
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219123910 10 mg
ICN19123925 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219123910 Supplier MP Biomedicals Inc Supplier No. 0219123910

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

α-Amylase from Porcine pancreas ≥700 u/mg protein, Opaque, White Suspension

  • It is used to hydrolyze α bonds of α-linked polysaccharides, such as starch and glycogen.
  • It has been used in various plant studies, such as metabolism studies in Arabidopsis.
  • It is isolated from porcine pancreas is a glycoprotein.
  • It is a single polypeptide chain of ∽475 residues containing two SH groups and four disulfide bridges and a tightly bound Ca2+ necessary for stability. Chloride ions are necessary for activity and stability.

Specifications

Color Opaque White
pH The pH range for activity is 5.5 to 8.0, with the pH optimum at 7.
Quantity 10 mg
Synonym β-N-acetylglucosaminidase porcine placenta
Form Suspension