missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Aldehyde Dehydrogenase, Potassium Activated
Description
Activity: 1U/mg solid (approx. 5U/mg protein). ADH Less than 0.1%. Unit Definition: One unit causes the oxidation of one micromole of acetaldehyde to acetic acid per minute at pH 8.0 and 25°C in the presence of potassium ions and thiols. Freeze dried stabilized powder.
Specifications
Specifications
| Concentration | ≈5 U/mg Protein, ≥1 U/mg Solid |
| Color | Off-White |
| pH | 9 |
| For Use With (Application) | Oxidizes a number of Aliphatic and Aromatic Aldehydes |
| Content And Storage | -20°C |
| Source | Yeast |
| Quantity | 100 U |
| Product Type | Aldehyde Dehydrogenase |
| Molecular Weight (g/mol) | ≈200000 |
| Inhibitors | SH-reagents, such as N-ethylmaleimide, 2-iodobenzoic Acid, Iodoacetic Acid and Chloromercuribenzoic Acid, Heavy Metals (especially Cu2-) and Acetaldehyde (at high concentrations) |
| Show More |