missing translation for 'onlineSavingsMsg'
Learn More

Moxalactam sodium salt, MilliporeSigma™ Supelco™

Chemical structure: ß-lactam

$447.13 - $1467.16

Specifications

CAS 64953-12-4
Linear Formula C20H18N6 Na2O9 S
MDL Number MFCD03427565
Type Analytical Standard
Formula Weight 564.44
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111015073
View Documents
Merck Emd Millipore
M81581G
1 g
Each for $447.13
Only null left
 
111015074
View Documents
Merck Emd Millipore
M81585G
5 g
Each for $1,467.16
Only null left
 
Description

Description

Specifications

Specifications

64953-12-4
MFCD03427565
564.44
C20H18N6 Na2O9 S
Analytical Standard
Analytical Standard
Videos
SDS
Documents

Documents