missing translation for 'onlineSavingsMsg'
Learn More

Moxalactam sodium salt, MilliporeSigma™ Supelco™

Catalog No. 111015073
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015073 1 g
111015074 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015073 Supplier Merck Emd Millipore Supplier No. M81581G
Only null left

Chemical structure: ß-lactam

Specifications

CAS 64953-12-4
Linear Formula C20H18N6 Na2O9 S
MDL Number MFCD03427565
Type Analytical Standard
Quantity 1 g
Formula Weight 564.44
Grade Analytical Standard