missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IP-10 (CXCL10) ELISA Kit

ELISA

$890.00 - $6080.00

Specifications

Accession Number P17515
Assay Range 7.8 to 500 pg/mL
Assay Sensitivity 6.5 pg/mL
Conjugate HRP
Product Type ELISA
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to mouse IP-10
Biotin-Conjugate anti-mouse IP-10 polyclonal antibody
Streptavidin-HRP
Mouse IP-10 Standard lyophilized, 1000pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5018292
View Documents
Invitrogen™
BMS6018
96 Tests
Each for $890.00
Only null left
 
5018294
View Documents
Invitrogen™
BMS6018TEN
10 x 96 Tests
Each for $6,080.00
Only null left
 
Description

Description

The Mouse Inflammatory Protein 10/CXCL10 (Ms IP-10) ELISA quantitates Ms IP-10 in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IP-10. Principle of the method The Mouse IP-10 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.
Specifications

Specifications

P17515
6.5 pg/mL
ELISA
Mouse
Colorimetric Microplate Reader
C7,CRG-2,INP10,IP-10,IP10,Ifi10,Scyb10,gIP-10,mob-1
8.4
HRP
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Interferon-gamma-inducible protein 10, CXCL10
1 hrs 20 mins
7.8 to 500 pg/mL
HRP
Plasma, Serum, Supernatant
ELISA
15945
7.2%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
CXCL10 (IP-10)
3 hrs 10 mins
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'