Learn More
Invitrogen™ Mouse IP-10 (CXCL10) ELISA Kit
ELISA
Supplier: Invitrogen™ BMS6018TEN
Description
The Mouse Inflammatory Protein 10/CXCL10 (Ms IP-10) ELISA quantitates Ms IP-10 in mouse serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IP-10. Principle of the method The Mouse IP-10 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.Specifications
| P17515 | |
| 6.5 pg/mL | |
| ELISA | |
| Mouse | |
| Colorimetric Microplate Reader | |
| C7,CRG-2,INP10,IP-10,IP10,Ifi10,Scyb10,gIP-10,mob-1 | |
| 8.4 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| CXCL10 (IP-10) | |
| 3 hrs 10 mins |
| 7.8 to 500 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| ELISA | |
| 15945 | |
| 7.2% | |
| Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 10 x 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Interferon-gamma-inducible protein 10, CXCL10 | |
| 1 hrs 20 mins |