missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-α-Tubulin, MP Biomedicals™
Mouse Monoclonal Antibody
Supplier: MP Biomedicals Inc 08L100028
Description
- Mouse anti-α-Tubulin mAb offers high specificity and potency and consistency against α-Tubulin expressed by human, mouse, and rat species, which has been experimentally validated in Western blotting (WB), Immunohistochemistry (IHC), Immunoprecipitation (IP) and Immunofluorescence (IF) assays.
- Microtubules of the eukaryotic cytoskeleton perform essential and diverse functions and are composed of a heterodimer of alpha and beta tubulin. The genes encoding these microtubule constituents are part of the tubulin superfamily, which is composed of six distinct families.
- Genes from the alpha, beta, and gamma tubulin families are found in all eukaryotes. The alpha and beta tubulins represent the major components of microtubules, while gamma tubulin plays a critical role in the nucleation of microtubule assembly. There are multiple alpha and beta tubulin genes, and they are highly conserved among and between species. This gene encodes an alpha tubulin that is a highly conserved homolog of a rat testis-specific alpha tubulin. Alternatively, spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| α-Tubulin | |
| Monoclonal | |
| In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol; pH 7.3 to 7.4 | |
| Full-length recombinant protein of human α-Tubulin | |
| 100 μL | |
| Secondary | |
| Observed at 50 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Affinity Purification | |
| RUO | |
| Human, Mouse, Rat | |
| Purified |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.