missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Rat Kappa Light Chain Secondary Antibody, FITC

Catalog No. 501137041
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501137041 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501137041 Supplier Invitrogen™ Supplier No. A15426
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Secondary Antibody

Mouse anti-Rat Kappa Light Chain Secondary Antibody for ELISA, Flow, ICC/IF

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rat Kappa Light Chain
Applications ELISA, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone K4F5
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with <0.1% sodium azide; pH 7.4
Gene Alias Ig kappa chain, Igk, Ig-kappa
Host Species Mouse
Immunogen This mouse IgG1-kappa class-switch variant monoclonal antibody reacts with rat kappa light chains and can be used for identification and enumeration of rat kappa light chain+ cells by flow cytometry.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage 4°C, store in dark
Product Type Secondary Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less