missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgG2b Heavy Chain Secondary Antibody (MARG2b-8) [Janelia Fluor 525], R&D Systems™
Description
IgG2b Heavy Chain Monoclonal antibody specifically detects IgG2b Heavy Chain in Rat samples. It is validated for ELISA
Specifications
Specifications
| Antigen | IgG2b Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MARG2b-8 |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | IR863 rat IgG2b myeloma protein |
| Purification Method | Affinity purified |
| Quantity | 0.25 mL |
| Show More |