missing translation for 'onlineSavingsMsg'
Learn More

IgG2b γ2b chainRat anti-Rat, Cy5 , Secondary Antibody, MilliporeSigma™

Catalog No. MAB505SMI
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB505SMI 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB505SMI Supplier MilliporeSigma Supplier No. MAB505S

Rat Monoclonal Antibody

Mouse anti-Rat IgG2b γ2b chain Antibody, Cy5 conjugate detects level of Rat IgG2b γ2b chain and has been published and validated for use in ELISA and immunofluoresence.

Specifications

Antigen Mouse anti-Rat IgG2b γ2b chain
Applications ELISA, Immunofluorescence
Classification Monoclonal
Conjugate Cyanine5
Formulation Purified by affinity chromatography. Liquid in 1.0 mL PBS/NaN3.
Host Species Rat
Immunogen Prepared from rat IgG2b γ2b chain.
Purification Method Affinity chromatography
Quantity 0.1 mg
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Do not Freeze.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only