missing translation for 'onlineSavingsMsg'
Learn More

IgG2b γ2b chainMouse anti-Rat, Biotin , Secondary Antibody, MilliporeSigma™

Catalog No. MAB505BMI
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB505BMI 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB505BMI Supplier MilliporeSigma Supplier No. MAB505B

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Mouse anti-Rat IgG2b γ2b chain Antibody, biotin conjugate is an antibody against Rat IgG2b γ2b chain for use in ELISA, flow cytometry.

Specifications

Antigen Mouse anti-Rat IgG2b γ2b chain
Applications ELISA, Flow Cytometry
Classification Monoclonal
Conjugate Biotin
Formulation Purified by affinity chromatography. Liquid in 1.0 mL PBS/NaN3.
Host Species Mouse
Immunogen Prepared from rat IgG2b γ2b chain.
Purification Method Affinity chromatography
Quantity 0.5 mg
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Show More Show Less

For Research Use Only