missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgG2a Heavy Chain Secondary Antibody (MARG2a-1) [Janelia Fluor 669], R&D Systems™
Description
IgG2a Heavy Chain Monoclonal antibody specifically detects IgG2a Heavy Chain in Rat samples. It is validated for ELISA
Specifications
Specifications
| Antigen | IgG2a Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MARG2a-1 |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Purified rat IgG2a |
| Purification Method | Affinity purified |
| Quantity | 0.25 mL |
| Show More |