missing translation for 'onlineSavingsMsg'
Learn More

IgG2a γ2a chainMouse anti-Rat, Alkaline Phosphatase , Secondary Antibody, MilliporeSigma™

Catalog No. MAB504AMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB504AMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB504AMI Supplier MilliporeSigma Supplier No. MAB504A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Mouse anti-Rat IgG2a γ2a chain Antibody, Alkaline Phosphatase detects level of Rat IgG2a γ2a chain and has been published and validated for use in ELISA.

Specifications

Antigen Mouse anti-Rat IgG2a γ2a chain
Applications ELISA
Classification Monoclonal
Conjugate Alkaline Phosphatase (AP)
Formulation Purified by affinity chromatography. Liquid in 50mM Tris/1mM MgCl2/50% Glycerol, pH 8.0, containing 0.1% NaN3 as preservative.
Host Species Mouse
Immunogen Prepared from rat IgG2a γ2a chain.
Purification Method Affinity chromatography
Quantity 1 mL
Primary or Secondary Secondary
Target Species Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only