missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgG1 Secondary Antibody, NovaFluor™ Blue 760, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$267.00 - $588.00
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 2D1 |
| Concentration | 0.8 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H005T03B11A
|
Invitrogen™
H005T03B11A |
100 Tests |
Each for $588.00
|
|
|||||
|
H005T02B11A
|
Invitrogen™
H005T02B11A |
25 Tests |
Each for $267.00
|
|
|||||
Description
Rat IgG1 Monoclonal antibody specifically detects Rat IgG1 in Human samples. It is validated for Flow CytometrySpecifications
| CD45 | |
| 0.8 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity chromatography |
| 2D1 | |
| Flow Cytometry | |
| NovaFluor Blue 760 | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |