missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Rat IgE Secondary Antibody, HRP

Catalog No. PIMA516813
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516813 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516813 Supplier Invitrogen™ Supplier No. MA516813

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Secondary Antibody

Mouse anti Rat IgE antibody, clone MARE-1 recognizes rat epsilon heavy chain of immunoglobulin IgE and does not cross-react with other classes of rat immunoglobulin.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Rat IgE
Applications ELISA, Western Blot
Classification Monoclonal
Clone MARE-1
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 50% glycerol and 0.01% thimerosal
Host Species Mouse
Immunogen Rat IgE myeloma protein (IR162, IR410).
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less