missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgD Secondary Antibody (MARD-3) [Janelia Fluor 525], R&D Systems™
Description
IgD Monoclonal antibody specifically detects IgD in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | IgD |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | MARD-3 |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta |
| Host Species | Mouse |
| Immunogen | IR731 myeloma protein |
| Purification Method | Affinity purified |
| Show More |