missing translation for 'onlineSavingsMsg'
Learn More

Light chain Mouse anti-Rabbit, Biotin , Secondary Antibody, MilliporeSigma™

Catalog No. MAB201BMI
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB201BMI 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB201BMI Supplier MilliporeSigma Supplier No. MAB201B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Mouse anti-Rabbit light chain Antibody, biotin conjugate is an antibody against Rabbit light chain for use in ELISA, flow cytometry, western blot, immunohistochemistry.

Specifications

Antigen Mouse anti-Rabbit light chain
Applications ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Biotin
Formulation Isolated from ascites by chromatographic procedures. Liquid in 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 0.05% Sodium Azide. 15mg/mL BSA as stabilizer.
Host Species Mouse
Immunogen Prepared from rabbit IgG light chain.
Purification Method Ascites
Quantity 0.5 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Keep away from light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only