missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rabbit IgM (H+L) Secondary Antibody (NRBM) [Alexa Fluor™ Plus 680], R&D Systems™
Description
IgM (H+L) Monoclonal antibody specifically detects IgM (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | IgM (H+L) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | NRBM |
| Conjugate | Alexa Fluor Plus 680 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Rabbit IgM |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |