missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rabbit IgM (H+L) Secondary Antibody (NRBM) [Alexa Fluor™ Plus 555], R&D Systems™

Catalog No. RD10128097
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10128097 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10128097 Supplier Bio-Techne Supplier No. NBP268467AFP555

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgM (H+L) Monoclonal antibody specifically detects IgM (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications

Antigen IgM (H+L)
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone NRBM
Conjugate Alexa Fluor Plus 555
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Rabbit IgM
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less