missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rabbit IgG (H+L) Secondary Antibody [Janelia Fluor 669], R&D Systems™
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Dot Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Mouse anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed) was produced by repeated immunization with rabbit whole IgG molecule in mouse. |
| Purification Method | Multi-step |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |