missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Monkey IgG (H+L) Secondary Antibody

Catalog No. PISA510345
Change view
Click to view available options
Quantity:
100 μg
500 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PISA510345 100 μg
01671620 500 μg
PISA510344 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PISA510345 Supplier Invitrogen™ Supplier No. SA510345
Only null left

Mouse Monoclonal Secondary Antibody

SA1-10653 detects monkey IgG, and will not cross-react with human, bovine, equine or ovine IgG. SA1-10653 has been successfully used in ELISA applications. The SA1-10653 immunogen is purified Rhesus monkey IgG.

Anti-Monkey secondary antibodies are affinity-purified antibodies with well-characterized specificity for non-human primate (e.g. monkey) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Monkey IgG (H+L)
Applications ELISA
Classification Monoclonal
Clone 5C12.D4
Concentration 1.4 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Host Species Mouse
Immunogen Purified Rhesus monkey IgG.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Monkey
Content And Storage -20°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less