missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Monkey IgA Secondary Antibody, Biotin

Catalog No. PIMA516730
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516730 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516730 Supplier Invitrogen™ Supplier No. MA516730

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Secondary Antibody

This antibody recognizes monkey IgA and does not cross-react with IgG or IgM; it has been shown to detect secreted IgA in cat saliva and rhesus monkey jejunal washes.

Anti-Monkey secondary antibodies are affinity-purified antibodies with well-characterized specificity for non-human primate (e.g. monkey) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Monkey IgA
Applications ELISA
Classification Monoclonal
Clone IgA5-3B
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with <0.1% sodium azide
Host Species Mouse
Immunogen purified IgA from serum.
Purification Method Protein A
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less