missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Human Kappa Light Chain Secondary Antibody, PE

Catalog No. PIMA110389
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA110389 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA110389 Supplier Invitrogen™ Supplier No. MA110389
  • ONLINE EXCLUSIVE PRICE
    $626.98
    $364.50 / Each

    Save $262.48

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Secondary Antibody

This antibody reacts with kappa light chains (22.5 kDa) of immunoglobulins. This antibody will not cross-react with hamster, goat, guinea pig, rabbit or sheep.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human Kappa Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone A8B5
Concentration 0.035 mg/mL
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Host Species Mouse
Immunogen not available.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less