missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Human IgM Secondary Antibody

Catalog No. sa172512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SA172512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA172512 Supplier Invitrogen™ Supplier No. SA172512

Mouse Monoclonal Secondary Antibody

This antibody reacts with the IgM chain C region and does not cross-react with human IgG and recombinant human IgG1, IgG2, IgG3, IgG4, IgA or IgE. This can be used in an Indirect ELISA (1 μg/mL) or as a capture antibody in a sandwich ELISA (5 μg/mL).

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human IgM
Applications ELISA, Western Blot
Classification Monoclonal
Clone KT16
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene IGHM
Gene Accession No. P01871
Gene Symbols IGHM
Host Species Mouse
Immunogen Recombinant human IgM.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3507
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less