missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Human IgM Secondary Antibody

Numéro de catalogue. 01676768
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
01676768 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 01676768 Fournisseur Invitrogen™ Code fournisseur SA119257

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Secondary Antibody

This antibody reacts with μ-chain of human IgM, Fab - region localized epitope.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Human IgM
Applications ELISA
Classification Monoclonal
Clone MA2
Concentration 1 mg/mL
Conjugué Unconjugated
Formule TBS with 15mM sodium azide; pH 8
Espèces hôtes Mouse
Immunogène IgM from human serum.
Méthode de purification Sequential chromatography
Quantité 100 μg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Human
Contenu et stockage 4°C, do not freeze
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG2b
Afficher plus de résultats Afficher moins de résultats