missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgM Heavy Chain Secondary Antibody (47.2.E9) [Janelia Fluor 669], R&D Systems™
Description
IgM Heavy Chain Monoclonal antibody specifically detects IgM Heavy Chain in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | IgM Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 47.2.E9 |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain |
| Host Species | Mouse |
| Immunogen | Purified human IgM |
| Purification Method | Protein A purified |
| Show More |