missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgM Fc Secondary Antibody (CH2) [Janelia Fluor 669], R&D Systems™
Description
IgM Fc Monoclonal antibody specifically detects IgM Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | IgM Fc |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | CH2 |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Purified human IgM. |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |