missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Human IgG4 Fc Secondary Antibody, HRP

Catalog No. MA182201
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA182201 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA182201 Supplier Invitrogen™ Supplier No. MA182201
  • ONLINE EXCLUSIVE PRICE
    $2,012.50
    $1,170.00 / Each

    Save $842.50

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

missing translation for 'validMainframeMsg'

Mouse Monoclonal Secondary Antibody

MA1-82201 detects the Fc portion of human IgG4. No cross-reactivity is observed with human IgG1, IgG2, and IgG3 isotypes. A suggested histology positive control is tonsil tissue. This antibody reacts with Human, African green monkey, Cynomolgus monkey, Baboon, Chimpanzee and Rhesus Monkey IgG. Mouse anti Human IgG4 antibody, clone HP6025 recognizes the heavy chain of human IgG4, at an epitope in the Fc region. No cross-reactivity is observed with IgG1, IgG2, IgG3, IgM, IgA (Jefferis et al. 1985).

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human IgG4 Fc
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HP6025
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 0.01% thimerosal
Host Species Mouse
Immunogen Purified IgG4.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less