missing translation for 'onlineSavingsMsg'
Learn More

IgG4 Mouse anti-Human, Clone: HP6023, Secondary Antibody, MilliporeSigma™

Catalog No. MAB1312MI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1312MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1312MI Supplier MilliporeSigma Supplier No. MAB1312

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Mouse anti-Human IgG4 Antibody, clone HP6023, Fc is an antibody against Human IgG4 for use in ELISA, RIA and western blotting.

Specifications

Antigen Mouse anti-Human IgG4 Fc
Applications ELISA, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone HP6023
Conjugate Unconjugated
Formulation Purified immunoglobulin. 0.02MPB+ 0.25M NaCl , Ph=7.6, 0.1% soduim azide.
Gene Accession No. NM_001002273.1; NM_001002274.1; NM_001002275.1; NM_004001.3
Host Species Mouse
Quantity 500 μg
Primary or Secondary Secondary
Gene ID (Entrez) FCGR2B; CDw32; CD32B; IGFR2; CD32; Fc-gamma-RIIb; FCG2; FcRII-b; FCGR2
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only