missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG4 Fc Secondary Antibody (5C7cc), R&D Systems™
Description
IgG4 Fc Monoclonal specifically detects IgG4 Fc in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | IgG4 Fc |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 5C7 |
| Conjugate | Unconjugated |
| Dilution | ELISA assay dependent |
| Gene Symbols | IGHG4 |
| Host Species | Mouse |
| Immunogen | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG4 from human serum. |
| Purification Method | Protein A purified |
| Show More |
For Research Use Only