missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG3 Heavy Chain Secondary Antibody (5G12cc) [Janelia Fluor 669], R&D Systems™
Description
IgG3 Heavy Chain Monoclonal antibody specifically detects IgG3 Heavy Chain in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | IgG3 Heavy Chain |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 5G12cc |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG3 from human serum. |
| Purification Method | Protein A purified |
| Quantity | 0.2 mL |
| Show More |