missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Human IgG2 Fc Secondary Antibody (52G1) [Janelia Fluor 525], R&D Systems™

Catalog No. NB788168
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB788168 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB788168 Supplier Bio-Techne Supplier No. NB1107082JF525

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG2 Fc Monoclonal antibody specifically detects IgG2 Fc in Human samples. It is validated for ELISA

Specifications

Antigen IgG2 Fc
Applications ELISA
Classification Monoclonal
Clone 52G1
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human IgG2 from myeloma patient serum.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less