missing translation for 'onlineSavingsMsg'
Learn More

IgG1 Mouse anti-Human, Clone: HP6001, Secondary Antibody, MilliporeSigma™

Catalog No. MAB1307MI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1307MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1307MI Supplier MilliporeSigma Supplier No. MAB1307

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Detect Human IgG1 using this Mouse anti-Human IgG1 Antibody, clone HP6001, Fc validated for use in ELISA, RIA and western blotting.

Specifications

Antigen Mouse anti-Human IgG1
Applications ELISA, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone HP6001
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02MPhosphate buffer, 0.25MNaCl containing 0.1% sodium azide.
Gene Accession No. NM_001002273.1; NM_001002274.1; NM_001002275.1; NM_004001.3
Host Species Mouse
Quantity 500 μg
Primary or Secondary Secondary
Gene ID (Entrez) FCGR2B; CDw32; CD32B; IGFR2; CD32; Fc-gamma-RIIb; FCG2; FcRII-b; FCGR2
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only