missing translation for 'onlineSavingsMsg'
Learn More

IgG1 Mouse anti-Human, Clone: 8c/6-39, Secondary Antibody, MilliporeSigma™

Catalog No. CBL309MI
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL309MI 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL309MI Supplier MilliporeSigma Supplier No. CBL309

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Mouse anti-Human IgG1 Antibody, clone 8c/6-39, Fc detects level of Human IgG1 and has been published and validated for use in ELISA, immunohistochemistry.

Specifications

Antigen Mouse anti-Human IgG1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 8c/6-39
Conjugate Unconjugated
Formulation The monoclonal is presented in liquid in 0.01M phosphate buffered saline containing 0.1% sodium azide, pH7.1. We recommend that each laboratory determine an optimum working titre for use in its particular application.
Host Species Mouse
Quantity 0.5 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only