missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG Secondary Antibody (MR12) [Alexa Fluor™ Plus 555], R&D Systems™
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Immunohistochemistry
Specifications
Specifications
| Antigen | IgG |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | MR12 |
| Conjugate | Alexa Fluor Plus 555 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Host Species | Mouse |
| Immunogen | Immunoglobulin purified from rabbit serum |
| Purification Method | Protein A purified |
| Show More |