missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG Heavy Chain Secondary Antibody (4D2D9G8) [Janelia Fluor 525], R&D Systems™
Description
IgG Heavy Chain Monoclonal antibody specifically detects IgG Heavy Chain in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | IgG Heavy Chain |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 4D2D9G8 |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Immunoglobulin G, ImmunoglobulinG |
| Host Species | Mouse |
| Immunogen | Human IgG was isolated from human sera and purified by chromatography. |
| Purification Method | Protein G purified |
| Show More |