missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Human IgG (H+L) Secondary Antibody (4D2D9G8), PE/Atto594, R&D Systems™

Catalog No. NB018696
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018696 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB018696 Supplier Bio-Techne Supplier No. NBP151523PEATT594

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG (H+L) Monoclonal specifically detects IgG (H+L) in Human samples. It is validated for ELISA, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4D2D9G8
Conjugate PE/ATTO 594
Dilution Western Blot : 1:500 - 1:2000, ELISA : 1:10000
Formulation PBS
Host Species Mouse
Immunogen Human IgG was isolated from human sera and purified by chromatography.
Purification Method Protein G purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less