missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG (H+L) Secondary Antibody (4D2D9G8), Janelia Fluor 549, R&D Systems™
Description
IgG (H+L) Monoclonal specifically detects IgG (H+L) in Human samples. It is validated for ELISA, Western Blot.
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4D2D9G8 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot : 1:500 - 1:2000, ELISA : 1:10000 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Human IgG was isolated from human sera and purified by chromatography. |
| Purification Method | Protein G purified |
| Show More |