missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG Fc Secondary Antibody (EM-07), R&D Systems™
Description
IgG Fc Monoclonal specifically detects IgG Fc in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot.
Specifications
Specifications
| Antigen | IgG Fc |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | EM-07 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Flow Cytometry, ELISA 1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Host Species | Mouse |
| Immunogen | Fusion protein of human IgG Fc fragment. |
| Purification Method | Protein A purified |
| Show More |
For Research Use Only