missing translation for 'onlineSavingsMsg'
Learn More

IgG Mouse anti-Human, Clone: HP6045, Secondary Antibody, MilliporeSigma™

Catalog No. MAB1304MI
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1304MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1304MI Supplier MilliporeSigma Supplier No. MAB1304

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

This Mouse anti-Human IgG Antibody, Fd, clone HP6045 is validated for use in ELISA, RIA, western blotting for the detection of Human IgG.

Specifications

Antigen Mouse anti-Human IgG
Applications ELISA, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone HP6045
Conjugate Unconjugated
Formulation Chromatographically purified IgG: isolated using DEAE and hyroxylapatite ion exchange chromatography. Phosphate buffered saline: pH 7.4, no azide.
Host Species Mouse
Quantity 500 μg
Primary or Secondary Secondary
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only