missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Human IgG Fc Secondary Antibody (3D3cc) [Janelia Fluor 669], R&D Systems™

Catalog No. NB789274
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB789274 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB789274 Supplier Bio-Techne Supplier No. NB1108347JF669

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgG Fc Monoclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, Immunodiffusion, Sandwich ELISA

Specifications

Antigen IgG Fc
Applications Western Blot, Immunodiffusion, Sandwich ELISA
Classification Monoclonal
Clone 3D3cc
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG from human serum.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less