missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Mouse anti-Human IgG, Fc Fragment Specific (HP6043), Peroxidase, Clone: HP6043, Secondary Antibody,

Catalog No. 411550500UG
Change view
Click to view available options
Quantity:
500 μg
Conjugate:
Peroxidase
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
411550500UG 500 μg Peroxidase
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 411550500UG Supplier MilliporeSigma™ Supplier No. 411550500UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Specifically detects IgG, Fc Fragment Specific (HP6043) Clone: HP6043 in Human samples, and it is validated for Enzyme Immunoassay

The HP series of murine human Ig-specific monoclonal antibodies was produced from documented hybridoma cell lines that were developed at the Centers for Disease Control in Atlanta, Georgia. Laboratory analysis of each monoclonal antibody lot is performed in two stages during the production of ascites. First, the cell culture medium is analyzed for the presence and relative amount of human Ig-specific antibody by ELISA before injection of cells into mice. Second, collected ascites are analyzed for potency and antibody specificity using ELISA and isoelectric focusing (IEF), often in combination with an affinity immunoblot. For more information, the CALBIOCHEM™ technical monograph, The Human IgG Subclasses, is available free of charge.

Specifications

Antigen IgG, Fc Fragment Specific (HP6043)
Applications ELISA
Classification Monoclonal
Clone HP6043
Conjugate Peroxidase
Formulation In PBS, 10mg/mL BSA.
Host Species Mouse
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage −20°C
Isotype IgG
Show More Show Less

For Research Use Only