missing translation for 'onlineSavingsMsg'
Learn More

IgG Mouse anti-Human, Clone: MK1A6, Secondary Antibody, MilliporeSigma™

Catalog No. CBL101MI
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL101MI 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL101MI Supplier MilliporeSigma Supplier No. CBL101

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Mouse anti-Human IgG Antibody, flow cytometry, all subclasses, clone MK1A6 detects level of Human IgG and has been published and validated for use in ELISA, immunofluoresence, HI, immunohistochemistry.

Specifications

Antigen Mouse anti-Human IgG
Applications ELISA, Agglutination, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone MK1A6
Conjugate Unconjugated
Formulation The monoclonal was purified by protein A affinity chromatography and is presented as a liquid in phosphate buffered saline containing 10mM sodium azide.
Host Species Mouse
Immunogen Polyclonal human IgG
Quantity 0.5 mg
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C for up to 1 month −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only