missing translation for 'onlineSavingsMsg'
Learn More

Mouse Anti-Human IgG Antibody [2C12H6], ProSci

Catalog No. 16595157
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16595157 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16595157 Supplier ProSci Llc Supplier No. PM67270100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Control Antibody

Mouse Anti-Human IgG Antibody specifically detects Mouse Anti-Human IgG in Human samples. It is validated for ELISA, Western Blot, Immunohistochemistry

Specifications

Antigen Mouse Anti-Human IgG
Applications ELISA, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 2C12H6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunoblotting: 1/5,000 - 1/50,000, ELISA: 1/5,000 - 1/50,000, Immunohistochemistry: 1/100 - 1/1,000
Formulation PBS pH 7.2, 1% BSA, 0.02% Thimerosal
Host Species Mouse
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No cross reactivity with immunoglobulins from other species.
Target Species Human
Content And Storage Antibody can be stored at -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Antibody
Form Liquid
Isotype IgG2
Show More Show Less