missing translation for 'onlineSavingsMsg'
Learn More

IgA Mouse anti-Human, FITC, Clone: M24A, Secondary Antibody, MilliporeSigma™

Catalog No. CBL114FMI
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL114FMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL114FMI Supplier MilliporeSigma Supplier No. CBL114F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

This Mouse anti-Human IgA Antibody, clone M24A, heavy chain, FITC conjugated is validated for use in ELISA, flow cytometry, immunofluoresence, immunohistochemistry for the detection of Human IgA.

Specifications

Antigen Mouse anti-Human IgA
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone M24A
Conjugate FITC
Formulation The conjugate is supplied in phosphate buffered saline containing 10mM sodium azide and 1mg/mL bovine serum albumin. For flow cytometry we recommend using 10μL of conjugate per test.
Host Species Mouse
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Isotype IgG1
Show More Show Less

For Research Use Only