missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Human Ig Mix Secondary Antibody

Catalog No. PISA510349
Change view
Click to view available options
Quantity:
100 μg
500 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PISA510349 100 μg
01671615 500 μg
PISA510348 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PISA510349 Supplier Invitrogen™ Supplier No. SA510349

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Secondary Antibody

SA1-10648 detects human Immunoglobulin. SA1-10648 has been successfully used in ELISA applications. The SA1-10648 immunogen is purified human immunoglobulin.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Specifications

Antigen Human Ig Mix
Applications ELISA
Classification Monoclonal
Clone HIG10-101.1.19
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Host Species Mouse
Immunogen Purified human immunoglobulin.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less